Learn More
Tau Monoclonal Antibody (HT7), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MN1000B
Code : PN
Additional Details : Weight : 0.80000kg
Description
MN1000B targets Tau in ELISA, WB and IHC applications and shows reactivity with Bovine, and Human samples. The MN1000B immunogen is purified human Tau, epitope human Tau between residue 159 and 163 (numbering according to human Tau40), corresponding to the amino acid sequence PPGQK. MN1000B detects Tau which has a predicted molecular weight of approximately 79 kDa.
Tau Monoclonal antibody specifically detects Tau in Bovine, Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Tau | |
Monoclonal | |
0.1 mg/mL | |
PBS with no preservative | |
P10636, P29172 | |
MAPT | |
Purified human Tau, epitope human Tau between residue 159 and 163 (numbering according to human Tau40), corresponding to the amino acid sequence PPGQK. | |
50 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry | |
HT7 | |
Biotin | |
MAPT | |
AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
Mouse | |
Protein A | |
RUO | |
281296, 4137 | |
4° C | |
Liquid |