missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA116996
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: Hela whole cell extract, antigen standard for TARDBP (transient overexpression lysate).
TDP-43 Polyclonal antibody specifically detects TDP-43 in Chicken, Human, Mouse, Non-human Primate, X. laevis, Zebrafish samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TDP-43 | |
Polyclonal | |
Unconjugated | |
TARDBP | |
1190002A23Rik; ALS10; C85084; hypothetical protein; OTTHUMP00000002171; OTTHUMP00000002173; RCJMB04_5g9; RP4-635E18.2; TAR DNA binding protein; TAR DNA binding protein L homeolog; TAR DNA-binding protein 43; TAR DNA-binding protein-43; tardbp; tardbp.L; Tdp43; TDP-43; wu:fb77f02; wu:fc52g10; XELAEV_18035188mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230908, 23435, 325052, 380329, 419453 | |
4° C, do not freeze | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q13148, Q5ZLN5, Q921F2 | |
TARDBP, tardbp.L | |
Synthetic peptide corresponding to C terminal region within residues 350-414 of the human TARDBP protein. | |
100 μL | |
Primary | |
Chicken, Human, Mouse, Non-human Primate, X. laevis, Zebrafish | |
Antibody | |
IgG |