Learn More
TAPP1 Rat anti-Human, Clone: 376409, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4039
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450376
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 17-12-2019
Description
Description
TAPP1 Monoclonal antibody MAB4039 Specifically detects TAPP1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
TAPP1/PLEKHA1
376409
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PLEKHA1
E. coli-derived recombinant human TAPP1 Pro2-Val404 Accession # Q9HB21
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
59338
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9HB21
family A (phosphoinositide bindingspecific) member 1, PH domain-containing family A member 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 1, PLEKHA1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TAPP1 in direct ELISAs and Western blots.