Learn More
TAFA2/FAM19A2 Mouse anti-Human, Clone: 447815, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4179
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450446
  • £288.60 / 100µg
Estimated Shipment date
from Supplier 06-07-2020
Description
Description
TAFA2/FAM19A2 Monoclonal antibody MAB4179 Specifically detects TAFA2/FAM19A2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
TAFA2/FAM19A2
447815
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FAM19A2
E. coli-derived recombinant human TAFA2/FAM19A2 Ala31-His131 Accession # AAH28403
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
338811
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAH28403
Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, FAM19A2, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TAFA2/FAM19A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 3, 4, or 5 is observed.