missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF172 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528429
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TAF172 Polyclonal antibody specifically detects TAF172 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
TAF172 | |
Polyclonal | |
Unconjugated | |
BTAF1 | |
AI414500; AI447930; ATP-dependent helicase BTAF1; BTAF1; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, (Mot1 homolog, S. cerevisiae); BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1 homolog, S. cerevisiae); B-TFIID TATA-box binding protein associated factor 1; B-TFIID transcription factor-associated 170 kDa subunit; E430027O22Rik; MOT1; Mot1 homolog; RGD1564130; TAF(II)170; TAF170; TAF172; TAF-172; TAFII170; TATA-binding protein-associated factor 172; TBP-associated factor 170; TBP-associated factor 172 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9044 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O14981 | |
BTAF1 | |
Synthetic peptide corresponding to a region within amino acids 1787 and 1849 of Human TAF172 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |