missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517079
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
ADAM17 Polyclonal antibody specifically detects ADAM17 in Human, Non-human Primate samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
ADAM17 | |
Polyclonal | |
Unconjugated | |
ADAM17 | |
a disintegrin and metallopeptidase domain 17; a disintegrin and metalloprotease domain 17; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 17; a disintegrin and metalloproteinase domain 17 (tumor necrosis factor, alpha, converting enzyme); ADAM; ADAM 17; ADAM metallopeptidase domain 17; ADAM metallopeptidase domain 18; Adam17; ADAM18; ADAMs; CD156B; CD156b antigen; CSVP; Disintegrin and metalloproteinase domain-containing protein 17; metalloendopeptidases; NISBD; NISBD1; Snake venom-like protease; TACE; TNF-alpha convertase; TNF-alpha converting enzyme; TNF-alpha-converting enzyme; tumor necrosis factor, alpha, converting enzyme | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6868 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
90 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P78536 | |
ADAM17 | |
Synthetic peptide corresponding to amino acids surrounding Val200 of human TACE | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |