missing translation for 'onlineSavingsMsg'
Learn More

TACE/ADAM17 Ectodomain Mouse anti-Human, Clone: 111623, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB9302

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462006

  • £353.77 / 500µg


Estimated Shipment
21-12-2018
Add to basket

Description

Description

TACE/ADAM17 Ectodomain Monoclonal antibody MAB9302 Specifically detects TACE/ADAM17 Ectodomain in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

TACE/ADAM17
111623
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ADAM17
Insect ovarian cell line T. ni-derived recombinant human TACE/ADAM17, Pro18-Asn671 (predicted), Accession # P78536
Protein A or G purified from ascites
RUO
Primary
6868
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P78536
ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human TACE/ADAM17 Ectodomain in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human ADAM8, 9, 15, and recombinant mouse ADAM10 is observed.
Human
Videos

For Research Use Only