Learn More
TACE/ADAM17 Cytosolic Domain Mouse anti-Human, Clone: 136121, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2129
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459195
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
TACE/ADAM17 Cytosolic Domain Monoclonal antibody MAB2129 Specifically detects TACE/ADAM17 Cytosolic Domain in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
TACE/ADAM17
136121
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ADAM17
E. coli-derived recombinant human TACE/ADAM17 Asp695-Cys824 (predicted) Accession # P78536
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6868
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P78536
ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TACE/ADAM17 Cytosolic Domain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ADAM8, 9, 10, 15, or 33 is observed.