Learn More
TAAR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533068
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Baboon, Marmoset, Tamarin, Rabbit (100%); Orangutan, Monkey, Rat, Horse (93%); Mouse, Bat, Hamster, Panda, Pig (87%); Bovine, Dog, Elephant (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
TAAR5 Polyclonal antibody specifically detects TAAR5 in Human, Rabbit samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
TAAR5 | |
Polyclonal | |
Unconjugated | |
Taar5 | |
Gm227; hTaar5; MGC138414; MGC138416; mTaar5; PNR; Putative neurotransmitter receptor; RP11-295F4.5; TAAR5; TaR-5; trace amine associated receptor 5; trace amine receptor 5; trace amine-associated receptor 5; Trimethylamine receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100344162, 9038 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O14804 | |
Taar5 | |
Synthetic 15 amino acid peptide from N-terminal extracellular domain of human TAAR5. | |
50 μg | |
Primary | |
Human, Rabbit | |
Antibody | |
IgG |