missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534245
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Galago, Marmoset, Mouse, Rat, Hamster, Pig (100%) Bovine (92%) Horse (83%).
TAAR9 Polyclonal antibody specifically detects TAAR9 in Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TAAR9 | |
Polyclonal | |
Unconjugated | |
TAAR9 | |
Gm231; mTaar9; SNORF1 receptor; TA3; Taar9; TAR3; TaR-3; TAR9; taR-9; trace amine associated receptor 9 (gene/pseudogene); trace amine receptor 3; trace amine receptor 9; trace amine-associated receptor 9; trace-amine-associated receptor 9; Trar3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134860, 319107, 503558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q5QD04, Q923Y6, Q96RI9 | |
TAAR9 | |
Synthetic 12 amino acid peptide from 2nd extracellular domain of human TAAR9. | |
50 μg | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |