missing translation for 'onlineSavingsMsg'
Learn More

TAAR9, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA534246

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15385091

  • £294.00 / 50µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Percent identity with other species by BLAST analysis: Human (100%) Galago (88%).
Specifications

Specifications

TA3/TAAR9
Polyclonal
Unconjugated
TAAR9
Purified
TAAR9
Synthetic 16 amino acid peptide from C-terminus of human TAAR9.
Antigen affinity chromatography
Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
134860
Immunohistochemistry (Paraffin)
1mg/mL
PBS with <0.1% sodium azide
Q96RI9
TAAR9, SNORF1 receptor, TaR-3, TaR-9, TAR3, TAR9, Trace amine receptor 9, TRAR3, TA3, Trace amine receptor 3
Rabbit
IgG
50μg
Antibody
Polyclonal
Human
Videos