missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA120783
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-20783 detects TA3 in Human samples.
TAAR9 Polyclonal antibody specifically detects TAAR9 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TAAR9 | |
Polyclonal | |
Unconjugated | |
TAAR9 | |
Gm231; mTaar9; SNORF1 receptor; TA3; Taar9; TAR3; TaR-3; TAR9; taR-9; trace amine associated receptor 9 (gene/pseudogene); trace amine receptor 3; trace amine receptor 9; trace amine-associated receptor 9; trace-amine-associated receptor 9; Trar3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134860 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96RI9 | |
TAAR9 | |
Synthetic 16 amino acid peptide from C-terminus of human TA3 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |