Learn More
T-Lymphocytes Monoclonal Antibody (KEN-5), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180464
376.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
This clone recognizes rabbit (lapine) T-cells, including >90% thymocytes and approximately 40-45% mesenteric lymph node cells. It immunoprecipitates a 67 kDa protein from rabbit thymocytes. This antibody does not bind to rabbit CD5 transfectants and shows very limited binding to rabbit B-cells, which in adult rabbits have been demonstrated to express CD5. This clone may recognize an epitope that is dependant on glycosylation to maintain its structural configuration, which may explain why the antibody does not recognize recombinant rabbit CD5 produced in insect cells (which contain different glycans from those found in mammalian cells). For flow cytometry, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
T-Lymphocytes Monoclonal antibody specifically detects T-Lymphocytes in Rabbit samples. It is validated for Flow Cytometry.Specifications
T-Lymphocytes | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
Rabbit thymocytes. | |
100 Tests | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
KEN-5 | |
FITC | |
Mouse | |
Protein A | |
RUO | |
Rabbit | |
Antibody | |
IgG1 |