Learn More
Profilin-like Protein Goat anti-T. gondii, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3860
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428414
  • £342.51 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Profilin-like Protein Polyclonal antibody AF3860 Specifically detects Profilin-like Protein in T. gondii samples. It is validated for Western Blotting
Specifications
Specifications
Profilin-like Protein
LYOPH
Western Blot 0.1 ug/mL
AAX47289
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
E. coli-derived recombinant T. gondii Profilin-like Protein Ser2-Tyr163 Accession # AAX47289
Immunogen affinity purified
RUO
Primary
Detects T. gondii Profilin-like Protein in direct ELISAs and Western blots.
T. gondii