Learn More
Synuclein-alpha Mouse anti-Human, Clone: 189105, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1338
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438555
  • £420.32 / 500µg
Estimated Shipment
17-07-2020
Description
Description
Synuclein-alpha Monoclonal antibody MAB1338 Specifically detects Synuclein-alpha in Human samples. It is validated for Western Blotting
Specifications
Specifications
alpha-Synuclein
189105
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SNCA
E. coli-derived recombinant human alpha-Synuclein Met1-Ala140 Accession # P37840
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6622
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P37840
Lewy body) 4, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, SNCA, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human alpha-Synuclein in direct ELISAs and Western blots.