Learn More
anti-Synuclein, α Ab-4; Polyclonal; Millipore™ (Chemicon™)
Brand:  Merck Millipore AB5038
Code : Z1
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12778786
  • £398.67 / 50┬ÁL
Estimated Shipment
02-03-2018
Add to basket
Specifications
Specifications
Synuclein, α Ab-4
Serum
Human α-synuclein residues 111-131 coupled to KLH. The peptide sHuence is N-GILEDMPVDPDNEAYEMPSEEC-C.
RUO
Primary
Rat
Unconjugated
Rabbit
50μL
Neuroscience