Learn More
Syntaphilin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520528
Code : LD
Additional Details : Weight : 0.01000kg
Description
Despite its predicted molecular weight, Syntaphilin often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human brain tissue lysate. PA5-20528 can be used with blocking peptide PEP-0648.
Syntaphilin Polyclonal antibody specifically detects Syntaphilin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Syntaphilin | |
Polyclonal | |
Unconjugated | |
SNPH | |
6430515A01; AW045671; AW556958; AW743098; bA314N13.5; KIAA0374; mKIAA0374; Snph; Syntaphilin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
241727, 296267, 9751 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
B5DF41, O15079, Q80U23 | |
SNPH | |
An 18 amino acid peptide from near the amino terminus of human Syntaphilin. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |