Learn More
Synaptophysin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527286
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: human brain, mouse brain, rat brain, Mouse small intestine, LNCaP. Predicted reactivity: Mouse (94%), Rat (95%), Rhesus Monkey (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Synaptophysin Polyclonal antibody specifically detects Synaptophysin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Synaptophysin | |
Polyclonal | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20977, 24804, 6855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P07825, P08247, Q62277 | |
Syp | |
Recombinant fragment corresponding to a region within amino acids 170 and 313 of Human Synaptophysin | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |