Learn More
SAP97 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1044
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-044 detects synapse-associated protein 97 (SAP97) from rat and human tissues. This antibody does not detect other synapse-associated protein family members. PA1-044 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽140 kDa protein representing SAP97 from rat brain homogenate. The PA1-044 immunogen is a synthetic peptide corresponding to residues E(16) E Y R S K L S Q T E D R Q L R S S(33) of human SAP97. This sequence is completely conserved in rat and mouse. PA1-044 immunizing peptide (Cat. # PEP-022) is available for use in neutralization and control experiments.
SAP97 Polyclonal antibody specifically detects SAP97 in Human, Rat samples. It is validated for Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
SAP97 | |
Polyclonal | |
Unconjugated | |
DLG1 | |
B130052P05Rik; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large MAGUK scaffold protein 1; discs, large homolog 1; discs, large homolog 1 (Drosophila); discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; DLG1; Dlgh1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; mKIAA4187; presynaptic protein SAP97; SAP97; SAP-97; synapse-associated protein 97 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1739, 25252 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q12959, Q62696 | |
DLG1 | |
Synthetic peptide corresponding to residues E(16) E Y R S K L S Q T E D R Q L R S S(33) of human SAP97. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |