missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Monoclonal Antibody (8E2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511680
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-11680 targets Survivin in ICC/IF and IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-11680 immunogen is full length recombinant human survivin protein.
Survivin Monoclonal antibody specifically detects Survivin in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Survivin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
O15392, Q9JHY7 | |
Birc5 | |
Full length recombinant human survivin protein | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
8E2 | |
Unconjugated | |
Birc5 | |
AAC-11; AP14; Api4; apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; baculoviral IAP repeat-containing 5 (survivin); baculoviral IAP repeat-containing protein 5; BIRC 5; BIRC5; EPR 1; EPR-1; Iap4; inhibitor of apoptosis; survivin; survivin variant 3 alpha; survivin40; SVV; TIAP | |
Mouse | |
Protein G | |
RUO | |
332, 64041 | |
4° C | |
Liquid |