Learn More
SFTPB Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen SA172016
Code : LD
Additional Details : Weight : 0.01000kg
Description
SA1-72016 detects Surfactant Protein B in human, mouse, and rat samples. SA1-72016 has been successfully used in western blot procedures. The SA1-72016 immunogen is from Chicken immunized with SP-B purified from Infasurf.
SFTPB Polyclonal antibody specifically detects SFTPB in Bovine samples. It is validated for Western Blot.Specifications
SFTPB | |
Polyclonal | |
Unconjugated | |
SFTPB | |
18 kDa pulmonary-surfactant protein; 6 kDa protein; AI562151; P SFTB3; pot. surfactant-associated protein precursor; putative; preproprotein; PSP-B; Pulmonary surfactant protein 18; pulmonary surfactant protein B; pulmonary surfactant-associated protein B; pulmonary surfactant-associated protein B precursor; pulmonary surfactant-associated protein B; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SF-B; SFPB; SFTB3; SFTP3; Sftp-3; Sftpb; SMDP1; SP 18; SP-B; SPBS; surfactant associated protein B; surfactant protein B; surfactant protein-B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B | |
Chicken | |
IgY fraction | |
RUO | |
507398 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
20 mg/mL | |
PBS with 0.075% sodium azide; pH 7.2 | |
P15781 | |
SFTPB | |
SP-B purified from Infasurf. | |
10 mg | |
Primary | |
Bovine | |
Antibody | |
IgY |