missing translation for 'onlineSavingsMsg'
Learn More

Sulfatase Modifying Factor 2/SUMF2 Mouse anti-Human, Clone: 330312, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3594

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460156

  • £264.01 / 100µg


Estimated Shipment
02-10-2018
Add to basket

Description

Description

Sulfatase Modifying Factor 2/SUMF2 Monoclonal antibody MAB3594 Specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Sulfatase Modifying Factor 2/SUMF2
330312
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SUMF2
Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2, Gln26-Leu301, Accession # Q8NBJ7
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
25870
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q8NBJ7
C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. No cross-reactivity with recombinant human SUMF1 or recombinant mouse SUMF2 is observed.
Human
Videos

For Research Use Only