Learn More
Streptococcus pneumoniae Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA17259
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-7259 detects Streptococcus pneumoniae. This product cross-reacts with types 3, 4, 6, 7, 9, 14, 18, 19 and 23. The antiserum was not absorbed and may react with related microorganisms. PA1-7259 has been successfully used in immunofluorescence and ELISA procedures. The PA1-7259 immunogen is a whole cell blend of numerous serotypes. Short term storage, up to 6 months, is liquid at 4oC. For long term storage, aliquot and store at -20 oC.
Streptococcus pneumoniae Polyclonal antibody specifically detects Streptococcus pneumoniae in Bacteria samples. It is validated for ELISA, Immunocytochemistry.Specifications
Streptococcus pneumoniae | |
Polyclonal | |
Unconjugated | |
Pneumococcus; s pneumoniae; S. pneumoniae | |
Whole cell blend of numerous serotypes. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA, Immunocytochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |