Learn More
Streptococcus Group B Monoclonal Antibody (HSB11-238.6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110705
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10705 detects Streptococci Group B. MA1-10705 has been successfully used in ELISA applications. The MA1-10705 immunogen is UV-inactivated Group B Streptococcus cells, types Ia, Ib, Ic, II, and III.
Streptococcus Group B Monoclonal antibody specifically detects Streptococcus Group B in Bacteria samples. It is validated for ELISA.Specifications
Streptococcus Group B | |
Monoclonal | |
1.1 mg/mL | |
PBS with no preservative; pH 7.4 | |
UV-inactivated Group B Streptococcus cells, types Ia, Ib, Ic, II, and III. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
HSB11-238.6 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |