missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520403
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human spleen tissue lysate. PA5-20403 can be used with blocking peptide PEP-0520.
STEAP1 Polyclonal antibody specifically detects STEAP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
STEAP1 | |
Polyclonal | |
Unconjugated | |
STEAP1 | |
2410007B19Rik; metalloreductase STEAP1; PRSS24; six transmembrane epithelial antigen of the prostate 1; six-transmembrane epithelial antigen of prostate 1; Steap; Steap1; STEAP-1; tcag7.1170 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26872, 297738, 70358 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9CWR7, Q9UHE8 | |
STEAP1 | |
An 18 amino acid peptide from near the carboxy terminus of human STEAP1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |