Learn More
STAU2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530721
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG, mouse brain. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STAU2 Polyclonal antibody specifically detects STAU2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
STAU2 | |
Polyclonal | |
Unconjugated | |
Stau2 | |
39K2; 39K3; Double-stranded RNA-binding protein Staufen homolog 2; r-staufen protein; Stau2; staufen (RNA binding protein) homolog 2 (Drosophila); staufen 2; staufen double-stranded RNA binding protein 2; staufen homolog 2; staufen, RNA binding protein, homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27067, 29819 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.67 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8CJ67, Q9NUL3 | |
Stau2 | |
Recombinant fragment corresponding to a region within amino acids 245 and 539 of Human STAU2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |