missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518562
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
STAT3 Polyclonal antibody specifically detects STAT3 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
STAT3 | |
Polyclonal | |
Unconjugated | |
STAT3 | |
1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
20848, 6774 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P40763, P42227 | |
STAT3 | |
Synthetic peptide sequence (CRPESQEHPEADP) corresponding to the internal amino acids of STAT3 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |