Learn More
Stanniocalcin 2/STC-2 Rat anti-Human, Clone: 348217, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2830
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429725
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Stanniocalcin 2/STC-2 Monoclonal antibody MAB2830 Specifically detects Stanniocalcin 2/STC-2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Stanniocalcin 2/STC-2
348217
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
STC2
Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 2 Thr25-Arg302 Accession # O76061
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8614
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O76061
stanniocalcin 2, stanniocalcin-2, STC2, STC-2, STC-2Stanniocalcin-related protein, STC-related protein, STCRP
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Stanniocalcin 2 (STC-2) in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) STC-1 or rmSTC-2.