Learn More
ST8SIA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532215
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, Raji, THP-1, NCI-H929. Predicted reactivity: Mouse (95%), Rat (94%), Pig (97%), Chicken (89%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ST8SIA4 Polyclonal antibody specifically detects ST8SIA4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ST8SIA4 | |
Polyclonal | |
Unconjugated | |
St8sia4 | |
alpha-2,8-polysialyltransferase; alpha-2,8-polysialyltransferase IV; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8D; alpha-2,8-sialyltransferase ST8Sia IV; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase-like protein; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; N-glycan alpha 2,8-sialyltransferase; polysialyltransferase; polysialyltransferase-1; Pst; PST1; PST-1; Sia8D; sialyltransf; sialyltransferase 8 (alpha-2 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2, 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-sialyltransferase) D; sialyltransferase 8 (alpha-2, 8-sialytransferase) D; sialyltransferase 8D; Sialyltransferase St8Sia IV; sialytransferase St8Sia IV; siat 8D; SIAT8D; SIAT8-D; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7903 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q92187 | |
St8sia4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 359 of Human ST8SIA4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |