Learn More
ST8SIA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542727
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GSFYTHSPLT IQLTLSSHRC NLPPLSSEYT KDVGSKSQLV TANPSIIRQR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 92%
ST8SIA1 Polyclonal antibody specifically detects ST8SIA1 in Human samples. It is validated for Western Blot.Specifications
ST8SIA1 | |
Polyclonal | |
Unconjugated | |
ST8SIA1 | |
9330109E03Rik; alpha-2, 8-sialyltransferase; alpha-2, 8-sialytransferase; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 1; alpha-2,8-sialyltransferase 8A; alpha-N-acetylneuraminate: alpha-2,8-sialytransferase; alpha-N-acetylneuraminide alpha-2,8-sialyltransferase; disialoganglioside (GD3) synthase; ganglioside GD3 synthase; Ganglioside GT3 synthase; ganglioside-specific alpha-2,8-polysialyltransferase; GD3 synthase; GD3S; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) A; sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3 synthase) A; Sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-28-sialytransferase GD3 synthase); sialyltransferase 8 A; Sialyltransferase 8 A (alpha-N-acetylneuraminate: alpha-28-sialytransferase GD3 synthase) GenBank no: U53883; sialyltransferase 8A; sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, GD3 synthase); sialyltransferase St8Sia I; sialytransferase St8Sia I; Sia-T | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6489 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92185 | |
ST8SIA1 | |
Synthetic peptide directed towards the N-terminal of human ST8SIA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |