missing translation for 'onlineSavingsMsg'
Learn More

ST6 Gal Sialyltransferase 1/ST6GAL1 Mouse anti-Human, Clone: 396012, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB5924

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461136

  • £271.93 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

ST6 Gal Sialyltransferase 1/ST6GAL1 Monoclonal antibody MAB5924 Specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
396012
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ST6GAL1
Mouse myeloma cell line NS0-derived recombinant human ST6 Gal Sialytransferase 1/ST6GAL1, Lys27-Cys406, Accession # P15907
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6480
Sterile PBS to a final concentration of 0.5 mg/mL.
Western Blot
LYOPH
Western Blot 1 ug/mL
P15907
Alpha 2,6-ST, B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CD75, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1, SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human ST6 Gal Sialytransferase 1/ST6GAL1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ST6GAL2, rhST6GALNAC1, or rhST6GALNAC5 is observed.
Human
Videos

For Research Use Only