Learn More
ST3GAL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542911
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: FDPKVENNPD TLLVLVAFKA MDFHWIETIL SDKKRVRKGF WKQPPLIWDV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%; Sheep: 93%
ST3GAL4 Polyclonal antibody specifically detects ST3GAL4 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ST3GAL4 | |
Polyclonal | |
Unconjugated | |
St3gal4 | |
alpha 2,3-sialyltransferase IV; Alpha 2,3-ST 4; alpha-2,3-sialyltransferase; alpha2,3-sialyltransferase; alpha-2,3-sialyltransferase ST3Gal IV; alpha-3-N-acetylneuraminyltransferase; beta-galactoside alpha-2,3-sialyltransferase 4; CGS23; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4; FLJ11867; gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; glycosyltransferase; NANTA3; SAT3; SAT-3; sialyltransferase 4C; sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase); SIAT4; SIAT4C; SIAT4-C; ST3 beta-galactoside alpha-2,3-sialyltransferase 4; ST3Gal IV; ST3GAL4; ST3GalA.2; ST3GalIV; ST3GAL-IV; ST-4; STZ | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6484 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q11206 | |
St3gal4 | |
Synthetic peptide directed towards the middle region of human ST3GAL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |