missing translation for 'onlineSavingsMsg'
Learn More

ST2/IL-1 R4 Rat anti-Mouse, Clone: 245714, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1004

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428245

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ST2/IL-33R Monoclonal antibody specifically detects ST2/IL-33R in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

ST2/IL-33R
245714
Western Blot 1 ug/mL
P14719
DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse ST2/IL-33 R in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human ST2 and no cross-reactivity with recombinant mouse (rm) IL-1 R1 or rmIL-1 R2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Il1rl1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ST2/IL-33 R Ser27-Arg332 Accession # P14719
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9173
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only