missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA5 Monoclonal Antibody (8E11), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1144PE
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Immunofluorescence and flow cytometric analysis of MA1-144-PE shows positive staining of SSEA-5 in human embryonal carcinoma NCCIT cells.
SSEA5 Monoclonal antibody specifically detects SSEA5 in Human samples. It is validated for Flow Cytometry.Specifications
SSEA5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
8E11 | |
PE | |
SSEA-5 | |
Undifferentiated H9 human embryonic stem cells | |
100 μL | |
Primary | |
4° C, do not freeze | |
Liquid |