missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRXN1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519037
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology.
SRXN1 Polyclonal antibody specifically detects SRXN1 in Human samples. It is validated for Western Blot.Specifications
SRXN1 | |
Polyclonal | |
Unconjugated | |
SRXN1 | |
1700127B04Rik; Ab2-390; AI854065; AW488194; C20orf139; dJ850E9.2; neoplastic progression 3; neoplastic progression protein 3; Npn3; Srx; Srx1; SRXN1; sulfiredoxin 1; sulfiredoxin 1 homolog (S. cerevisiae); sulfiredoxin-1; TX01; YKL086W | |
Goat | |
Antigen affinity chromatography | |
RUO | |
140809 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BYN0 | |
SRXN1 | |
Synthetic peptide sequence (KVQSLVDTIREDPD) corresponding to the internal amino acids of SRXN1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |