missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541770
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: DSEKAKKPVR SMIETRGEKP KEKAKNSKKK GAKKEGSDGP LATSKPVPAE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 93%
SRPR Polyclonal antibody specifically detects SRPR in Human samples. It is validated for Western Blot.Specifications
SRPR | |
Polyclonal | |
Unconjugated | |
Srpra | |
1300011P19Rik; D11Mgi27; Docking protein alpha; DP; DP-alpha; MGC17355; MGC3650; MGC9571; signal recognition particle receptor (docking protein); signal recognition particle receptor ('docking protein'); signal recognition particle receptor subunit alpha; Sralpha; SR-alpha; SRP receptor alpha subunit; SRP receptor subunit alpha; SRP-alpha; Srpr; SRPRA; Unknown (protein for MGC:128907) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6734 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P08240 | |
Srpra | |
Synthetic peptide directed towards the N-terminal of human SRPR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |