Learn More
SREBP1 Monoclonal Antibody (2A4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511685
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-11685 targets SREBP-1 in WB applications and shows reactivity with Hamster and Human samples. The MA5-11685 immunogen is recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1.
SREBP1 Monoclonal antibody specifically detects SREBP1 in Hamster, Human samples. It is validated for Western Blot.Specifications
SREBP1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
P36956, Q60416 | |
SREBF1 | |
Recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1 | |
500 μL | |
Primary | |
Hamster, Human | |
Antibody | |
IgG1 κ |
Western Blot | |
2A4 | |
Unconjugated | |
SREBF1 | |
ADD1; ADD-1; Adipocyte determination- and differentiation-dependent factor 1; bHLHd1; Class D basic helix-loop-helix protein 1; D630008H06; I79_010155; nt-SREBP1; Processed sterol regulatory element-binding protein 1; SREBF1; SREBP1; SREBP-1; SREBP1a; SREBP-1a; SREBP1c; SREBP-1c; sterol regulatory element binding protein 1; sterol regulatory element binding protein-1; sterol regulatory element binding transcription factor 1; sterol regulatory element binding-protein 1; sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1 | |
Mouse | |
Protein A/G | |
RUO | |
100689018, 6720 | |
4° C | |
Liquid |