missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA116803
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: mouse liver lysate or protein.Specifications
SR-BI | |
Polyclonal | |
Unconjugated | |
SCARB1 | |
Purified | |
SCARB1 | |
Synthetic peptide corresponding to the C-terminal residues 400-509 of mouse SR-BI. | |
Antigen affinity chromatography | |
Store at 4° C, do not freeze | |
Primary | |
949 |
Western Blot | |
3mg/mL | |
tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8WTV0, Q61009, P97943 | |
SCARB1, SR-B1, SR-BI, Scavenger Receptor-BI, Scavenger Receptor-B1 | |
Goat | |
IgG | |
100μL | |
Antibody | |
Polyclonal | |
Rodent |