missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERLIN2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518413
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology.
ERLIN2 Polyclonal antibody specifically detects ERLIN2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ERLIN2 | |
Polyclonal | |
Unconjugated | |
ERLIN2 | |
BC036333; C87251; C8orf2; EMBL:AAI68668.1, ECO:0000312; EMBL:AAI68668.1}; endoplasmic reticulum lipid raft-associated protein 2; endoplasmic reticulum lipid raft-associated protein 2 {ECO:0000250; ER lipid raft associated 2; Erlin2; erlin-2; erlin-2 {ECO:0000250; erlin2 {ECO:0000312; HGNC:1356; MGC87072; NET32; RGD:1309010}; RGD1309010; SPFH domain family, member 2; SPFH domain-containing protein 2; SPFH domain-containing protein 2 {ECO:0000250; Spfh2; SPG18; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2 {ECO:0000250; UniProtKB:Q8BFZ9, ECO:0000312; UniProtKB:Q8BFZ9}; UNQ2441; UNQ2441/PRO5003/PRO9924 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11160 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O94905 | |
ERLIN2 | |
Synthetic peptide sequence (EDEPLETATKEN) corresponding to the C-terminus amino acids of ERLIN2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |