Learn More
SPARC-like 1/SPARCL1 Rat anti-Mouse, Clone: 348905, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2836
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449725
  • £320.46 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
SPARC-like 1/SPARCL1 Monoclonal antibody MAB2836 Specifically detects SPARC-like 1/SPARCL1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
SPARC-like 1/SPARCL1
348905
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SPARCL1
Mouse myeloma cell line NS0-derived recombinant mouse SPARC-like 1/SPARCL1 Ile17-Phe650 Accession # P70663
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8404
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P70663
hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC like 1, SPARCL1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse SPARC-like 1/SPARCL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SPARC or recombinant human SPARC-like 1 is observed.