Learn More
SPA17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544107
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MSIPFSNTHY RIPQGFGNLL EGLTREILRE QPDNIPAFAA AYFESLLEKR Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 79%
SPA17 Polyclonal antibody specifically detects SPA17 in Human samples. It is validated for Western Blot.Specifications
SPA17 | |
Polyclonal | |
Unconjugated | |
SPA17 | |
band 34; cancer/testis antigen 22; CT22; SP17; SP17 protein; SP17-1; SPA17; Sperm autoantigenic protein 17; sperm protein 17; Sperm surface protein Sp17; Unknown (protein for MGC:127855); zgc:153721 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
53340 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15506 | |
SPA17 | |
Synthetic peptide directed towards the N-terminal of human SPA17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |