missing translation for 'onlineSavingsMsg'
Learn More

SOST/Sclerostin Rat anti-Mouse, Clone: 248121, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1589

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458755

  • £353.77 / 500µg


Estimated Shipment
03-12-2018
Add to basket

Description

Description

SOST/Sclerostin Monoclonal antibody MAB1589 Specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

SOST/Sclerostin
248121
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SOST
Mouse myeloma cell line NS0-derived recombinant mouse SOST, Gln24-Tyr211, Accession # AAI37834
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
50964
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAI37834
sclerostin, VBCHsclerosteosis
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse SOST in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human SOST.
Mouse
Videos

For Research Use Only