Learn More
SNX11 Monoclonal Antibody (2G1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA521788
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MGFWCRMSEN QEQEEVITVR VQDPRVQNEG SWNSYVDYKI FLHTNSKAFT AKTSCVRRRY REFVWLRKQL QRNAGLVPVP ELPGKSTFFG TSDEFIEKRR QGLQHFLEKV LQSVVLLSDS QLHLFLQSQL SVPEIEACVQ GRSTMTVSDA ILRYAMSNCG WAQEERQSSS HLAKGDQPKS CCFLPRSGRR SSPSPPPSEE KDHLEVWAPV VDSEVPSLES PTLPPLSSPL CCDFGRPKEG TSTLQSVRRA VGGDHAVPLD PGQLETVLEK
SNX11 Monoclonal antibody specifically detects SNX11 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
SNX11 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9Y5W9 | |
SNX11 | |
SNX11 (AAH00768, 1 a.a. ∽ 270 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
2G1 | |
Unconjugated | |
SNX11 | |
4933439F10Rik; A930041K09Rik; AI117694; MGC111019; SNX11; sorting nexin 11; sorting nexin-11 | |
Mouse | |
Affinity chromatography | |
RUO | |
29916 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |