missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOHLH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530658
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SOHLH2 Polyclonal antibody specifically detects SOHLH2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SOHLH2 | |
Polyclonal | |
Unconjugated | |
SOHLH2 | |
4933406N12Rik; AI413378; bHLHe81; RGD1304752; Sohlh2; SPATA28; spermatogenesis and oogenesis specific basic helix-loop-helix 2; spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 2; spermatogenesis associated 28; TEB1; testicular secretory protein Li 50; testicular secretory protein Li 51; Tohlh2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54937 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NX45 | |
SOHLH2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 228 of Human SOHLH2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |