Learn More
Soggy-1/DkkL1 Mouse anti-Human, Clone: 202509, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1549
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448725
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 10-07-2020
Description
Description
Soggy-1/DkkL1 Monoclonal antibody MAB1549 Specifically detects Soggy-1/DkkL1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Soggy-1/DkkL1
202509
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DKKL1
Mouse myeloma cell line NS0-derived recombinant human Soggy-1 Ala31-Leu242 Accession # Q9UK85.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27120
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9UK85.1
Cancer/testis antigen 34, CT34dickkopf-like 1 (soggy), dickkopf-like 1, DkkL1, Protein soggy-1, SGY, SGY1, SGY-1dickkopf-like protein 1, SGY1soggy, soggy-1, Soggy1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Soggy-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Soggy-1.