Learn More
ATP1A1 Monoclonal Antibody (M8-P1-A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA3929
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA3-929 detects sodium/potassium ATPase alpha 1 in sheep, canine, rat, mouse, human and porcine tissues. This antibody does not detect the alpha 1 isoform in Xenopus and chicken. MA3-929 does not detect the alpha 2 or alpha 3 isoform in rat. MA3-929 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, flow cytometry, and immunohistochemistry procedures. By Western blot, this antibody detects an ∽110 kDa protein representing alpha 1 sodium/potassium ATPase from canine kidney extract. Immunohistochemical staining of sodium/potassium ATPase alpha 1 with MA3-929 yields a pattern consistent with plasma membrane localization. The MA3-929 antigen is sheep kidney alpha 1 sodium/potassium ATPase. This antibody recognizes an epitope between amino acid residues 496-506 of lamb kidney sodium/potassium ATPase.
ATP1A1 Monoclonal antibody specifically detects ATP1A1 in Canine, Human, Mouse, Ovine, Porcine, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
ATP1A1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P04074, P05023, P05024, P06685, P50997, Q8VDN2 | |
ATP1A1 | |
Lamb kidney alpha 1 sodium/potassium ATPase. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Ovine, Porcine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
M8-P1-A3 | |
Unconjugated | |
ATP1A1 | |
(Na+ + K+)-ATPase; (Na+, K+)-ATPase alpha-subunit; alpha subunit of sodium potassium ATPase; atp1a1; atp1a1-a; atp1a1a.4; atp1a1-b; ATP1B; Atpa-1; ATPase; ATPase Na+/K+ transporting subunit alpha 1; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; BC010319; K-ATPase; MGC1798; Na(+)/K(+) ATPase alpha-1 subunit; Na, K-ATPase alpha-1 subunit; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na/K ATPase alpha 1 isoform; Na/K ATPase alpha 1 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ ATPase; Na+/K+ ATPase 1; Na+/K+ ATPase Alpha; Na+/K+ -ATPase alpha 1 subunit; Na+/K+ transporting alpha 1 polypeptide; Na+K+ ATPase; Na+K+ATPase; Nkaa1b; RP4-655J12.1; sarcolemmal sodium/potassium-ATPase alpha 1; Sodium Potassium ATPase; sodium pump 1; sodium pump subunit alpha-1; sodium/potassium-ATPase alpha-subunit 1; sodium/po | |
Mouse | |
Protein A | |
RUO | |
11928, 24211, 397481, 403992, 443381, 476 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |