missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519708
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken, cow and fruit fly (Drosophila melanogaster) based on sequence homology.
SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.Specifications
SNAP25 | |
Polyclonal | |
Unconjugated | |
P60880, P60879, P60881 | |
SNAP25 | |
Synthetic peptide conjugated to KLH derived from within residues 100 - 200 of Human SNAP25. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.1 mg/mL | |
SNAP25 | |
FLJ23079, RIC-4, RIC4, SEC9, SNAP, SNAP-25, bA416N4.2, dJ1068F16.2, SUP; resistance to inhibitors of cholinesterase 4 homolog; super protein; synaptosomal-associated 25 kDa protein; synaptosomal-associated protein 25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20614, 25012, 6616 | |
-20°C or -80°C if preferred | |
Liquid |