Learn More
SMURF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530900
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (96%), Xenopus laevis (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SMURF2 Polyclonal antibody specifically detects SMURF2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SMURF2 | |
Polyclonal | |
Unconjugated | |
Smurf2 | |
2810411E22Rik; AI558114; AI649275; E3 ubiquitin ligase SMURF2; E3 ubiquitin-protein ligase SMURF2; HECT-type E3 ubiquitin transferase SMURF2; hSMURF2; RGD1310067; SMAD specific E3 ubiquitin protein ligase 2; SMAD ubiquitination regulatory factor 2; SMAD-specific E3 ubiquitin-protein ligase 2; SMURF2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64750 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.82 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9HAU4 | |
Smurf2 | |
Recombinant fragment corresponding to a region within amino acids 306 and 549 of Human SMURF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |