Learn More
SLITRK5 Mouse anti-Human, Clone: 310516, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2587
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439555
  • £249.49 / 100µg
Estimated Shipment
28-06-2019
Description
Description
SLITRK5 Monoclonal antibody MAB2587 Specifically detects SLITRK5 in Human samples. It is validated for Western Blotting
Specifications
Specifications
SLITRK5
310516
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SLITRK5
Mouse myeloma cell line NS0-derived recombinant human SLITRK5 Glu39-Val665 Accession # O94991
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
26050
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O94991
bA364G4.2, FLJ58374, KIAA0918, leucine-rich repeat-containing protein 11, LRRC11, SLIT and NTRK-like family, member 5, SLIT and NTRK-like protein 5
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human SLITRK5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SLITRK1, 2, 3, or 4 is observed.