Learn More
SLITRK2 Mouse anti-Human, Clone: 312420, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2657
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439605
  • £249.49 / 100µg
Estimated Shipment
27-06-2019
Description
Description
SLITRK2 Monoclonal antibody MAB2657 Specifically detects SLITRK2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
SLITRK2
312420
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SLITRK2
Mouse myeloma cell line NS0-derived recombinant human SLITRK2 Thr19-Pro617 Accession # Q9H156
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84631
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9H156
CXorf2, SLITL1, SLITRK2 SLIT and NTRK-like family, member 2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human SLITRK2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) SLITRK1, rhSLITRK4, or rhSLITRK5 is observed.